missing translation for 'onlineSavingsMsg'
Learn More

Pergolide Mesilate, Mikromol™

Product Code. 18063350
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 18063350

Brand: Mikromol MM3400.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pergolide Mesilate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pergolide mesilate
CAS 66104-23-2
Analyte or Component Names Pergolide mesilate
Shipping Condition Room Temperature
Molecular Formula C19 H26 N2 S . C H4 O3 S
Certifications/Compliance ISO 17025
Synonym Pergolide mesilate, (6aR,9R,10aR)-9-[(Methylsulfanyl)methyl]-7-propyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline monomethanesulfonate, Indolo[4,3-fg]quinoline, ergoline deriv., Celance, LY 127809, NSC 319773, Pergolide mesylate, Pergolide methanesulfonate, Permax
InChI Formula InChI=1S/C19H26N2S.CH4O3S/c1-3-7-21-11-13(12-22-2)8-16-15-5-4-6-17-19(15)14(10-20-17)9-18(16)21;1-5(2,3)4/h4-6,10,13,16,18,20H,3,7-9,11-12H2,1-2H3;1H3,(H,2,3,4)/t13-,16-,18-;/m1./s1
SMILES CCCN1C[C@H](CSC)C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34.CS(=O)(=O)O
IUPAC Name (6aR,9R,10aR)-9-(methylsulfanylmethyl)-7-propyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline;methanesulfonic acid
Molecular Weight (g/mol) 410.59
Quantity 250 mg
Formula Weight 410.1698 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.