accessibility menu, dialog, popup

UserName

CAS RN 6025-53-2

CH3OHHNNNNN(R)O(R)OH(S)HO(R)HO(E)

CAS RN 6025-53-2

IUPAC Name: (2R,3R,4S,5R)-2-(6-{[(2E)-4-hydroxy-3-methylbut-2-en-1-yl]amino}-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: trans-zeatin riboside
Molecular Weight (g/mol): 351.36
Molecular Formula: C15H21N5O5
InChi Key: GOSWTRUMMSCNCW-HNNGNKQASA-N
SMILES: C\C(CO)=C/CNC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC=N1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)