Learn More
trans-Zeatin-riboside
Description
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
trans-Zeatin-riboside is used in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general.
Solubility
Soluble in water, ethanol, and methanol.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Specifications
| Color | White |
| Quantity | 50 mg |
| Beilstein | 5460894 |
| Solubility Information | Soluble in water,ethanol,and methanol. |
| Formula Weight | 351.36 |
| Physical Form | Powder |
| Chemical Name or Material | trans-Zeatin-riboside |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.