missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatinriboside, MedChemExpress

Product Code. 16490469
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490469 10 mg 10mg
16475447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490469 Supplier Medchem Express Supplier No. HYW011151

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress trans-Zeatinriboside is a type of cytokinin precursor, acts as a major long-distance signalling form in xylem vessels, regulates leaf size and meristem activity-related traits.

trans-Zeatinriboside is a type of cytokinin precursor, acts as a major long-distance signalling form in xylem vessels, regulates leaf size and meristem activity-related traits.

Specifications

Chemical Name or Material trans-Zeatinriboside
CAS 6025-53-2
Color White
Purity Grade Notes Research
Molecular Formula C15H21N5O5
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (284.61 mM)
SMILES C/C(CO)=C\CNC1=C(N=CN2[C@H]3[C@H](O)[C@H](O)[C@H](O3)CO)C2=NC=N1
Molecular Weight (g/mol) 351.36
Formula Weight 351.36
Percent Purity 99.51%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.