missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin-riboside, 97%

Product Code. 10489481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10489481 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10489481 Supplier Thermo Scientific Acros Supplier No. 337200500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6025-53-2 | C15H21N5O5 | 351.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6025-53-2
Melting Point 179°C to 180°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C15H21N5O5
MDL Number MFCD00036809
Quantity 50 mg
Synonym zeatin riboside, trans-zeatin riboside, trans-zeatin-riboside, ribosylzeatin, trans-zeatinriboside, 9-ribosyl-trans-zeatin, unii-7lg4d082a9, n-4-hydroxy-3-methyl-2-butenyl adenosine, e-n-4-hydroxy-3-methyl-2-butenyl adenosine, 9-beta-d-ribosyl-trans-zeatin
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
InChI Key GOSWTRUMMSCNCW-HNNGNKQASA-N
SMILES CC(=CCNC1=NC=NC2=C1N=CN2C3C(C(C(O3)CO)O)O)CO
IUPAC Name (2R,3S,4R,5R)-2-(hydroxymethyl)-5-[6-[[(E)-4-hydroxy-3-methylbut-2-enyl]amino]purin-9-yl]oxolane-3,4-diol
Molecular Weight (g/mol) 351.36
PubChem CID 6440982
ChEBI CHEBI:71693
Formula Weight 351.36
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material trans-Zeatin-riboside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.