missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Zeatin Riboside
Description
Naturally occurring cytokinin riboside.
Naturally occurring cytokinin riboside. Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence. Chromatographically purified and recrystallized. Plant cell culture tested. Mixed isomers: trans-isomer 97%, cis-isomer >2%.
Specifications
Specifications
| CAS | 6025-53-2 |
| Quantity | 1 g |
| Molecular Formula | C15H21N5O5 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?