accessibility menu, dialog, popup

UserName

H3CH3C(S)CH3HO(S)(S)(S)(S)OH(R)H3C(S)(S)H3C

CAS RN 516-72-3

IUPAC Name: (1S,3aS,3bS,7S,9aR,9bS,11aS)-1-[(2S)-2-hydroxy-6-methylheptan-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms: 20 α-hydroxycholesterol 20R-hydroxycholesterol
Molecular Weight (g/mol): 402.66
Molecular Formula: C27H46O2
InChi Key: MCKLJFJEQRYRQT-APGJSSKUSA-N
SMILES: CC(C)CCC[C@](C)(O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results