missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 20(S)-Hydroxycholesterol
GREENER_CHOICE

Product Code. 18096032
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18096032 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18096032 Supplier Tocris Bioscience™ Supplier No. 4474/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allosteric activator of Hedgehog (Hh) signaling; induces Smo accumulation

Allosteric activator of the Hedgehog signaling pathway Smoothened (Smo) oncoprotein; binds at a site distinct from the canonical cyclopamine binding site. Activates Hedgehog (Hh) signaling (EC50 ∽ 3μM for induction of Hh reporter gene transcription in in NIH 3T3 cells). Induces Smo accumulation in primary cilia; also exhibits osteogenic activity and activates the liver X receptor (LXR).
TRUSTED_SUSTAINABILITY

Specifications

CAS 516-72-3
Quantity 10 mg
Molecular Formula C27H46O2
Formula Weight 402.65
Purity >95%
Molecular Weight (g/mol) 402.663
Chemical Name or Material 20(S)-Hydroxycholesterol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.