missing translation for 'onlineSavingsMsg'
Learn More

20(S)-Hydroxycholesterol, MedChemExpress

Product Code. 16475088
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475088 10 mg 10mg
16407817 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16475088 Supplier Medchem Express Supplier No. HY12316

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 20(S)-hydroxyCholesterol (20α-Hydroxycholesterol) is an allosteric activator of the oncoprotein smoothened (Smo) that activates the hedgehog (Hh) signaling pathway with an EC50 of 3 μM in a gene transcription reporter assay using NIH3T3 cells.

20(S)-hydroxyCholesterol (20α-Hydroxycholesterol) is an allosteric activator of the oncoprotein smoothened (Smo) that activates the hedgehog (Hh) signaling pathway with an EC50 of 3 μM in a gene transcription reporter assay using NIH3T3 cells.

Specifications

Chemical Name or Material 20(S)-Hydroxycholesterol
CAS 516-72-3
Color White
Purity Grade Notes Research
Molecular Formula C27H46O2
Quantity 10 mg
Synonym 20α-Hydroxycholesterol
Solubility Information DMSO : 30 mg/mL (74.51 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@]1([C@@]2([H])[C@@](C)(O)CCCC(C)C)[C@](CC2)([H])[C@@](CC=C3[C@@]4(CC[C@H](O)C3)C)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 402.65
Formula Weight 402.65
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.