CAS RN 446859-33-2
CAS RN 446859-33-2
IUPAC Name:
2-[5-(6-methylpyridin-2-yl)-1H-pyrazol-4-yl]-1,5-naphthyridine
Synonyms:
2-[3-(6-methylpyridin-2-yl)-2H-pyrazol-4-yl]-1,5-naphthyridine
Molecular Weight (g/mol):
287.33
Molecular Formula:
C17H13N5
InChi Key:
LBPKYPYHDKKRFS-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(=N1)C1=C(C=NN1)C1=CC=C2N=CC=CC2=N1
1
–
7
of
7
results
1
Tocris Bioscience™ RepSox
CAS: 446859-33-2 Molecular Formula: C17H13N5
2
Alk 5 Inhibitor II, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Alk 5 Inhibitor II, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Tocris Bioscience™ RepSox
RepSox synthesized to Ancillary Material Grade
5
Alk 5 Inhibitor II, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
6
7
RepSox, MedChemExpress
MedChemExpress RepSox (E-616452) is a potent and selective of the TGFβR-1/ALK5 inhibitor which inhibits ALK5 autophosphorylation with an IC50 of 4 nM.