missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alk 5 Inhibitor II, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Alk 5 Inhibitor II |
| CAS | 446859-33-2 |
| Molecular Formula | C17H13N5 |
| Quantity | 50 mg |
| Synonym | 2-[3-(6-Methyl-2-pyridinyl)-1H-pyrazol-4-yl]-1,5-naphthyridine, E 616452, RepSox, SJN 2511 |
| InChI Formula | InChI=1S/C17H13N5/c1-11-4-2-5-16(20-11)17-12(10-19-22-17)13-7-8-14-15(21-13)6-3-9-18-14/h2-10H,1H3,(H,19,22) |
| SMILES | Cc1cccc(n1)c2n[nH]cc2c3ccc4ncccc4n3 |
| IUPAC Name | 2-[5-(6-methylpyridin-2-yl)-1H-pyrazol-4-yl]-1,5-naphthyridine |
| Molecular Weight (g/mol) | 287.32 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction