missing translation for 'onlineSavingsMsg'
Learn More

RepSox, MedChemExpress

Product Code. p-200100690
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485478

Brand: Medchem Express HY13012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RepSox (E-616452) is a potent and selective of the TGFβR-1/ALK5 inhibitor which inhibits ALK5 autophosphorylation with an IC50 of 4 nM.

RepSox (E-616452) is a potent and selective of the TGFβR-1/ALK5 inhibitor which inhibits ALK5 autophosphorylation with an IC50 of 4 nM.

Specifications

Chemical Name or Material RepSox
CAS 446859-33-2
Color Light Brown
Purity Grade Notes Research
Molecular Formula C17H13N5
Quantity 10 mg
Synonym E-616452 SJN 2511
Solubility Information DMSO : 33.33 mg/mL (116.00 mM; Need ultrasonic)
SMILES CC1=NC(C2=NNC=C2C3=NC4=CC=CN=C4C=C3)=CC=C1
Molecular Weight (g/mol) 287.32
Formula Weight 287.32
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.