missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RepSox
GREENER_CHOICE

Product Code. 18034202
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18034202

Brand: Tocris Bioscience™ 3742/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective inhibitor of TGF-βRI

RepSox is a potent and selective inhibitor of the TGF-β type I receptor/ALK5 (IC50 values are 4 and 23 nM for TGF-β type I receptor autophosphorylation and binding respectively). Selective for TGF-β type I receptor over a range of kinases, including p38 MAPK, JNK1 and GSK3 (IC50 > 16 μM). Enhances the efficiency of cellular reprogramming; replaces Sox2 by inducing Nanog expression.

RepSox synthesized to Ancillary Material Grade also available.

For more information about how RepSox may be used, see our protocols: Highly Efficient Generation of CiPSCs from MEFs, Generation of β cells from hPSCs, Reprogramming MEFs into Skeletal Muscle Progenitors .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446859-33-2
Molecular Formula C17H13N5

Specifications

Quantity 10 mg
Formula Weight 287.32
Purity >99%
Molecular Weight (g/mol) 287.326
Chemical Name or Material RepSox
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.