accessibility menu, dialog, popup

UserName

CH3CH3NOH3COCl

CAS RN 15972-60-8

IUPAC Name: 2-chloro-N-(2,6-diethylphenyl)-N-(methoxymethyl)acetamide
Synonyms: LAZO caswell no. 011 lasagrin alochlor chimiclor alachlore alachlor alanox alanex pillarzo
Molecular Weight (g/mol): 269.77
Molecular Formula: C14H20ClNO2
InChi Key: XCSGPAVHZFQHGE-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(CC)=C1N(COC)C(=O)CCl
Certifications/Compliance 
  • (8)
  • (1)
Solvent or Matrix 
  • (3)
  • (1)
  • (4)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Concentration 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (1)
  • (8)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)