missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 508.1 Pesticide Mixture Modified 100 μg/mL in Ethyl acetate, Dr. Ehrenstorfer

Product Code. 30315518
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315518 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315518 Supplier Dr. Ehrenstorfer Supplier No. DREA50000534EA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alachlor∣Atrazine∣Butachlor∣Chlorobenzilate∣Chloroneb∣Chlorothalonil∣Chlorthal-dimethyl∣Cis-Chlordane (Alpha Isomer)∣Cyanazine∣Etridiazole∣Hexachlorobenzene∣Hexachlorocyclopentadiene∣Metolachlor∣Metribuzin∣Permethrin∣Propachlor∣Simazine∣Trans-Chlordane (Gamma Isomer)∣Trifluralin
CAS 15972-60-8, 1912-24-9, 23184-66-9, 510-15-6, 2675-77-6, 1897-45-6
Analyte or Component Names Alachlor∣Atrazine∣Butachlor∣Chlorobenzilate∣Chloroneb∣Chlorothalonil∣Chlorthal-dimethyl∣Cis-Chlordane (Alpha Isomer)∣Cyanazine∣Etridiazole∣Hexachlorobenzene∣Hexachlorocyclopentadiene∣Metolachlor∣Metribuzin∣Permethrin∣Propachlor∣Simazine∣Trans-Chlordane (Gamma Isomer)∣Trifluralin
Shipping Condition +20°C
Certifications/Compliance ISO/IEC 17025
Solvent or Matrix Ethyl Acetate
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Alachlor 100 μg/mL∣Atrazine 100 μg/mL∣Butachlor 100 μg/mL∣Chlorobenzilate 100 μg/mL∣Chloroneb 100 μg/mL∣Chlorothalonil 100 μg/mL∣Chlorthal-dimethyl 100 μg/mL∣Cis-Chlordane (Alpha Isomer) 100 μg/mL∣Cyanazine 100 μg/mL∣Etridiazole 100 μg/mL∣Hexachlorobenzene 100 μg/mL∣Hexachlorocyclopentadiene 100 μg/mL∣Metolachlor 100 μg/mL∣Metribuzin 100 μg/mL∣Permethrin 100 μg/mL∣Propachlor 100 μg/mL∣Simazine 100 μg/mL∣Trans-Chlordane (Gamma Isomer) 100 μg/mL∣Trifluralin 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.