missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrogen/Phosphorus Pesticide Mixture 925 100 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Alachlor∣Ametryn∣Atraton∣Atrazine∣Bromacil∣Butachlor∣Butylate∣Chlorpropham∣Chlorpyrifos∣Cyanazine∣Cycloate∣Dichlorvos∣Diphenamid∣EPTC∣Ethoprophos∣Fenarimol∣Fluridone∣Hexazinone∣Metolachlor∣Mevinphos∣MGK 264∣Molinate∣Napropamide∣Norflurazon∣Paraoxon-methyl∣Pebulate∣Prometon∣Prometryn∣Propachlor∣Propazine∣Propyzamide∣Simetryn∣Tebuthiuron∣Terbacil∣Terbutryn∣Triadimefon∣Tricyclazole∣Trifluralin∣Vernolate |
| CAS | 15972-60-8, 834-12-8, 1610-17-9, 1912-24-9, 314-40-9, 23184-66-9 |
| Analyte or Component Names | Alachlor∣Ametryn∣Atraton∣Atrazine∣Bromacil∣Butachlor∣Butylate∣Chlorpropham∣Chlorpyrifos∣Cyanazine∣Cycloate∣Dichlorvos∣Diphenamid∣EPTC∣Ethoprophos∣Fenarimol∣Fluridone∣Hexazinone∣Metolachlor∣Mevinphos∣MGK 264∣Molinate∣Napropamide∣Norflurazon∣Paraoxon-methyl∣Pebulate∣Prometon∣Prometryn∣Propachlor∣Propazine∣Propyzamide∣Simetryn∣Tebuthiuron∣Terbacil∣Terbutryn∣Triadimefon∣Tricyclazole∣Trifluralin∣Vernolate |
| Shipping Condition | Ice pack (-20°C) |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Alachlor 100 μg/mL∣Ametryn 100 μg/mL∣Atraton 100 μg/mL∣Atrazine 100 μg/mL∣Bromacil 100 μg/mL∣Butachlor 100 μg/mL∣Butylate 100 μg/mL∣Chlorpropham 100 μg/mL∣Chlorpyrifos 100 μg/mL∣Cyanazine 100 μg/mL∣Cycloate 100 μg/mL∣Dichlorvos 100 μg/mL∣Diphenamid 100 μg/mL∣EPTC 100 μg/mL∣Ethoprophos 100 μg/mL∣Fenarimol 100 μg/mL∣Fluridone 100 μg/mL∣Hexazinone 100 μg/mL∣Metolachlor 100 μg/mL∣Mevinphos 100 μg/mL∣MGK 264 100 μg/mL∣Molinate 100 μg/mL∣Napropamide 100 μg/mL∣Norflurazon 100 μg/mL∣Paraoxon-methyl 100 μg/mL∣Pebulate 100 μg/mL∣Prometon 100 μg/mL∣Prometryn 100 μg/mL∣Propachlor 100 μg/mL∣Propazine 100 μg/mL∣Propyzamide 100 μg/mL∣Simetryn 100 μg/mL∣Tebuthiuron 100 μg/mL∣Terbacil 100 μg/mL∣Terbutryn 100 μg/mL∣Triadimefon 100 μg/mL∣Tricyclazole 100 μg/mL∣Trifluralin 100 μg/mL∣Vernolate 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?