missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 505 Organohalide Pesticide Mixture 388 200 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30317286
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317286 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317286 Supplier Dr. Ehrenstorfer Supplier No. DREA50000388ME
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alachlor∣Aldrin∣Atrazine∣Cis-Chlordane (Alpha Isomer)∣cis-Nonachlor∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Hexachlorocyclopentadiene∣Methoxychlor∣Simazine∣Trans-Chlordane (Gamma Isomer)∣trans-Nonachlor
CAS 15972-60-8, 309-00-2, 1912-24-9, 5103-71-9, 5103-73-1, 60-57-1
Analyte or Component Names Alachlor∣Aldrin∣Atrazine∣Cis-Chlordane (Alpha Isomer)∣cis-Nonachlor∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Hexachlorocyclopentadiene∣Methoxychlor∣Simazine∣Trans-Chlordane (Gamma Isomer)∣trans-Nonachlor
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Alachlor 200 μg/mL∣Aldrin 200 μg/mL∣Atrazine 200 μg/mL∣Cis-Chlordane (Alpha Isomer) 200 μg/mL∣cis-Nonachlor 200 μg/mL∣Dieldrin 200 μg/mL∣Endrin 200 μg/mL∣gamma-HCH 200 μg/mL∣Heptachlor 200 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 200 μg/mL∣Hexachlorobenzene 200 μg/mL∣Hexachlorocyclopentadiene 200 μg/mL∣Methoxychlor 200 μg/mL∣Simazine 200 μg/mL∣Trans-Chlordane (Gamma Isomer) 200 μg/mL∣trans-Nonachlor 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.