missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organohalide Pesticide Mix 2 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Alachlor∣Aldrin∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Methoxychlor |
| CAS | 15972-60-8, 309-00-2, 60-57-1, 72-20-8, 58-89-9, 76-44-8 |
| Analyte or Component Names | Alachlor∣Aldrin∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Methoxychlor |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solution Type | Mixture |
| Solvent or Matrix | Methanol |
| Industry Type | Environmental Testing |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Endrin 99.94 μg/mL∣gamma-HCH 99.94 μg/mL∣Aldrin 99.95 μg/mL∣Heptachlor 99.95 μg/mL∣Dieldrin 99.98 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 100 μg/mL∣Alachlor 100.02 μg/mL∣Hexachlorobenzene 100.02 μg/mL∣Heptachlor-endo-epoxide (trans-, isomer A) 100.05 μg/mL∣Methoxychlor 100.23 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?