CAS RN 146368-11-8
CAS RN 146368-11-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
CY5, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
CY5, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
CY5, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
MedChemExpress CY5
MedChemExpress Cy5 (Sulfo-Cyanine5) is a reactive dye for the labeling of amino-groups in peptides, proteins, and oligonucleotides.