missing translation for 'onlineSavingsMsg'
Learn More

CY5, TRC

Product Code. 30364658
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30364658

Brand: Toronto Research Chemicals TRCC9513105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(1E,3E,5E)-5-[1-(5-carboxypentyl)-3,3-dimethyl-5-sulfoindol-2-ylidene]penta-1,3-dienyl]-1-ethyl-3,3-dimethylindol-1-ium-5-sulfonate
CAS 146368-11-8
Molecular Formula C33 H40 N2 O8 S2
Quantity 5 mg
Synonym CY5, 2-(5-(1-(5-Carboxypentyl)-3,3-dimethyl-5-sulfoindolin-2-ylidene)penta-1,3-dien-1-yl)-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulfonate, 2-((1E,3E,5E)-5-(1-(5-Carboxypentyl)-3,3-dimethyl-5-sulfoindolin-2-ylidene)penta-1,3-dien-1-yl)-1-ethyl-3,3-dimethyl-3H-indol-1-ium-5-sulfonate, 2-[5-[1-(5-Carboxypentyl)-1,3-dihydro-3,3-dimethyl-5-sulfo-2H-indol-2-ylidene]-1,3-pentadienyl]-1-ethyl-3,3-dimethyl-5-sulfo-3H-indolium
InChI Formula InChI=1S/C33H40N2O8S2/c1-6-34-27-18-16-23(44(38,39)40)21-25(27)32(2,3)29(34)13-9-7-10-14-30-33(4,5)26-22-24(45(41,42)43)17-19-28(26)35(30)20-12-8-11-15-31(36)37/h7,9-10,13-14,16-19,21-22H,6,8,11-12,15,20H2,1-5H3,(H2-,36,37,38,39,40,41,42,43)
SMILES CC[N+]1=C(\C=C\C=C\C=C/2\N(CCCCCC(=O)O)c3ccc(cc3C2(C)C)S(=O)(=O)O)C(C)(C)c4cc(ccc14)S(=O)(=O)[O-]
IUPAC Name 2-[(1E,3E,5E)-5-[1-(5-carboxypentyl)-3,3-dimethyl-5-sulfoindol-2-ylidene]penta-1,3-dienyl]-1-ethyl-3,3-dimethylindol-1-ium-5-sulfonate
Molecular Weight (g/mol) 656.809
Formula Weight 656.223
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.