missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress CY5

Product Code. 16481178
Encompass_Preferred
Click to view available options
:
5 mg
This item is not returnable. View return policy

Product Code. 16481178

Brand: MedChemExpress HYD0821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cy5 (Sulfo-Cyanine5) is a reactive dye for the labeling of amino-groups in peptides, proteins, and oligonucleotides.

This dye requires small amount of organic co-solvent (such as DMF or DMSO) to be used in labeling reaction. This reagent is ideal for very cost-efficient labeling of soluble proteins, as well as all kinds of peptides and oligonucleotides. This reagent also works well in organic solvents for small molecule labeling. Excitation (nm):649, Emission (nm): 670.

Specifications

Quantity 5 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 656.81 g/mol
Solubility DMSO : 250 mg/mL (380.63 mM; Need ultrasonic) ∣ H2O : 25 mg/mL (38.06 mM; Need ultrasonic)
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Target HBV
CAS 146368-11-8
Synonym Sulfo-Cyanine5
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.