CAS RN 1224606-06-7
CAS RN 1224606-06-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ DLin-MC3-DMA
Component of lipid nanoparticles (LNPs); ionizable cationic lipid
2
D-Lin-MC3-DMA, MedChemExpress
MedChemExpress D-Lin-MC3-DMA, an ionizable cationic lipid, is a potent siRNA delivery vehicle.