missing translation for 'onlineSavingsMsg'
Learn More

D-Lin-MC3-DMA, MedChemExpress

Product Code. 16497317
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497317 5 mg 5mg
16473156 25 mg 25mg
16444608 10 mg 10mg
16440907 50 mg 50mg
16323237 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497317 Supplier Medchem Express Supplier No. HY112251

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress D-Lin-MC3-DMA, an ionizable cationic lipid, is a potent siRNA delivery vehicle.

D-Lin-MC3-DMA, an ionizable cationic lipid, is a potent siRNA delivery vehicle.

Specifications

Chemical Name or Material D-Lin-MC3-DMA
Physical Form Liquid
CAS 1224606-06-7
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C43H79NO2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (155.74 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (155.74 mM; Need ultrasonic)
SMILES O=C(OC(CCCCCCCC/C=C\C/C=C\CCCCC)CCCCCCCC/C=C\C/C=C\CCCCC)CCCN(C)C
Molecular Weight (g/mol) 642.09
Formula Weight 642.09
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.