missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DLin-MC3-DMA

Product Code. 18706045
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18706045 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18706045 Supplier Tocris Bioscience™ Supplier No. 7946/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Component of lipid nanoparticles (LNPs); ionizable cationic lipid

DLin-MC3-DMA is an ionizable cationic lipid (pKa = 6.44), used in combination with other lipids in the formation of lipid nanoparticles (LNPs) for delivering cargos such as oligonucleotides into living cells. DLin-MC3-DMA can protect negatively charged cargos and facilitate their cytosolic transport.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6 Z,9 Z,28 Z,31 Z)-Heptatriaconta-6,9,28,31-tetraen-19-yl 4-(dimethylamino)butanoate
CAS 1224606-06-7
Quantity 25 mg
Target General Laboratory Research Reagent
Molecular Formula C43H79NO2
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.