missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF217 Recombinant Rabbit Monoclonal Antibody (24GB4185)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen ZNF217
Clone 24GB4185
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275674
GREENER_CHOICE
View Documents
Invitrogen™
MA557327
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275145
View Documents
Invitrogen™
MA557328
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF217 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Binds to the promoters of target genes and functions as repressor. Promotes cell proliferation and antagonizes cell death. Promotes phosphorylation of AKT1 at 'Ser-473'.
Specifications

Specifications

ZNF217
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Rat
ZNF217
ZABC1; Zinc finger protein 217; ZNF217
ZNF217
Primary
Affinity Chromatography
24GB4185
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75362
7764
A synthesized peptide derived from human ZNF217 (aa 1000-1048).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.