missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF217 Recombinant Rabbit Monoclonal Antibody (24GB4185)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557328
This item is not returnable.
View return policy
Description
ZNF217 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Binds to the promoters of target genes and functions as repressor. Promotes cell proliferation and antagonizes cell death. Promotes phosphorylation of AKT1 at 'Ser-473'.
Specifications
| ZNF217 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ZNF217 | |
| ZABC1; Zinc finger protein 217; ZNF217 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7764 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4185 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75362 | |
| ZNF217 | |
| A synthesized peptide derived from human ZNF217 (aa 1000-1048). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction