missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH Zirconium Oxide Comfort Grinding Jars
Use these grinding jars with Retsch™ ball mills. Retsch™ Zirconium Oxide Comfort Grinding Jars feature an O-type sealing for safe operation.
£4510.00 - £6935.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10380722
|
RETSCH
01.462.0187 |
125 mL |
£4510.00
Each |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||||
10671083
|
RETSCH
01.462.0219 |
250 mL |
£5970.00
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
10161002
|
RETSCH
01.462.0227 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Available in a variety of sizes
- Optional aeration covers
- Pressure tight safety closure devices
Specifications
Comfort Grinding Jar | |
Zirconium Oxide |