missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Z-Guggulsterone
Broad spectrum steroid receptor ligand. More active isomer of guggulsterone (Cat. No. 2013)
£168.21 - £639.65
Specifications
Purity | >98% |
---|---|
Product Type | Z-Guggulsterone |
Molecular Weight (g/mol) | 312.45 |
Inhibitors | Z-Guggulsterone |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18095222
|
Tocris Bioscience™
3570/50 |
50 mg |
£639.65
50mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18095212
|
Tocris Bioscience™
3570/10 |
10 mg |
£168.21
10mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Broad spectrum steroid receptor ligand; mineralocorticoid, progesterone and glucocorticoid receptor antagonist (Ki values are 37, 224 and 252 nM respectively) and weak androgen receptor agonist (Ki = 315 nM). Induces apoptosis in prostate cancer cells and inhibits angiogenesis via suppression of the VEGF-VEGFR2-Akt signaling pathway. Exhibits antilipidemic activity via antagonism of the farnesoid X receptor (FXR) and displays antiseptic, antirheumatic and anti-inflammatory activity in vivo. More active isomer of guggulsterone (Cat. No. 2013).Specifications
>98% | |
Z-Guggulsterone | |
312.45 | |
Z-Guggulsterone |
For Research Use Only