Learn More
Tocris Bioscience™ Z-Guggulsterone
Broad spectrum steroid receptor ligand. More active isomer of guggulsterone (Cat. No. 2013)
Brand: Tocris Bioscience™ 3570/10
Additional Details : Weight : 0.00970kg
Description
Z-Guggulsterone is a broad spectrum steroid receptor ligand; mineralocorticoid, progesterone and glucocorticoid receptor antagonist (Ki values are 37, 224 and 252 nM respectively) and weak androgen receptor agonist (Ki = 315 nM). Induces apoptosis in prostate cancer cells and inhibits angiogenesis via suppression of the VEGF-VEGFR2-Akt signaling pathway. Exhibits antilipidemic activity via antagonism of the farnesoid X receptor (FXR) and displays antiseptic, antirheumatic and anti-inflammatory activity in vivo. More active isomer of guggulsterone (Cat. No. 2013).Specifications
>98% | |
Z-Guggulsterone | |
Z-Guggulsterone |
10 mg | |
312.45 |
For Research Use Only