missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT5B Recombinant Rabbit Monoclonal Antibody (24GB2690)

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen WNT5B
Clone 24GB2690
Concentration 0.76 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275752
View Documents
Invitrogen™
MA557321
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276015
View Documents
Invitrogen™
MA557322
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

WNT5B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 94% and 80% amino acid identity to the mouse Wnt5b protein and the human WNT5A protein, respectively. Alternative splicing of this gene generates 2 transcript variants.
Specifications

Specifications

WNT5B
0.76 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P22726, Q9H1J7
22419, 282582, 81029
A synthesized peptide derived from human Wnt5b (aa 300-359).
Antibody
24GB2690
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
WNT5B
AW545702; protein Wnt-5b; wingless-related MMTV integration site 5B; wingless-type MMTV integration site family member 5B; wingless-type MMTV integration site family, member 5B; Wnt family member 5B; WNT5B; Wnt-5b; WNT-5B protein
WNT5B
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.