missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT5B Recombinant Rabbit Monoclonal Antibody (24GB2690)
Description
WNT5B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 94% and 80% amino acid identity to the mouse Wnt5b protein and the human WNT5A protein, respectively. Alternative splicing of this gene generates 2 transcript variants.
Specifications
Specifications
| Antigen | WNT5B |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 24GB2690 |
| Concentration | 0.76 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | WNT5B |
| Gene Accession No. | P22726, Q9H1J7 |
| Gene Alias | AW545702; protein Wnt-5b; wingless-related MMTV integration site 5B; wingless-type MMTV integration site family member 5B; wingless-type MMTV integration site family, member 5B; Wnt family member 5B; WNT5B; Wnt-5b; WNT-5B protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?