Learn More
Invitrogen™ WNT5B Recombinant Rabbit Monoclonal Antibody (24GB2690)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557322
 View more versions of this product
Product Code. 30276015
  • £126.00 / 20µL
Description
WNT5B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 94% and 80% amino acid identity to the mouse Wnt5b protein and the human WNT5A protein, respectively. Alternative splicing of this gene generates 2 transcript variants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WNT5B
Recombinant Monoclonal
0.76 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P22726, Q9H1J7
WNT5B
A synthesized peptide derived from human Wnt5b (aa 300-359).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2690
Unconjugated
WNT5B
AW545702; protein Wnt-5b; wingless-related MMTV integration site 5B; wingless-type MMTV integration site family member 5B; wingless-type MMTV integration site family, member 5B; Wnt family member 5B; WNT5B; Wnt-5b; WNT-5B protein
Rabbit
Affinity chromatography
RUO
22419, 282582, 81029
-20°C