missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | WIPI2 |
|---|---|
| Concentration | 300 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16846245
|
Proteintech
28820-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16836245
|
Proteintech
28820-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The WIPI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WIPI2. This antibody recognizes human, mouse antigen. The WIPI2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| WIPI2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
| 300 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 26100 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title