Learn More
WIPI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28820-1-AP-150UL
 View more versions of this product
Product Code. 16836245
  • £349.00 / 150µL
Description
The WIPI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WIPI2. This antibody recognizes human, mouse antigen. The WIPI2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
WIPI2
Polyclonal
Unconjugated
150 μL
26100
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
300 μg/mL
Rabbit
Primary
Human, Mouse