Learn More
WIPI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28820-1-AP-20UL
Description
The WIPI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WIPI2. This antibody recognizes human, mouse antigen. The WIPI2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| WIPI2 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 26100 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |