missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Trypsinizing Flasks

Deep fluted sides enhance agitation

Brand:  DWK Life Sciences 355442

Code : 03

Additional Details : Weight : 0.13700kg

 View more versions of this product

Product Code. 11581275

  • £75.10 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket

Description

Description

Deep fluted sides enhance agitation

  • Manufactured from low-extractable borosilicate glass
  • Excellent quality and low extractables
  • Deep-skirted rubber-lined screw caps
  • Available graduated or ungraduated

Specifications

Specifications

Style: Erlenmeyer Trypsinizing
Borosilicate Glass
430
Yes
24 mm
35 mL
Used for forming cell suspensions through Trypsin digestion of connective Proteins
Autoclavable
Screw
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers