missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Wee1, GST Tag Recombinant Protein
WEE1 is classified as a Ser/Thr kinase by sequence analysis.
£434.00 - £6890.00
Specifications
Accession Number | NP_003381.1 |
---|---|
Gene Alias | WEE1 |
KinaseFamily | WEE Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11593547
|
Invitrogen™
PV3817 |
10 μg |
£434.00
10µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10148672
|
Invitrogen™
PV4072 |
1mg |
£6890.00
1mg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478573
|
Invitrogen™
PR7373A |
100μg |
£1578.00
100µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
WEE1 has been shown to inactivate multiple CDKs by tyrosine phosphorylation.
Specifications
NP_003381.1 | |
WEE Kinase Family | |
Recombinant | |
Baculovirus |
WEE1 | |
Human | |
GST Tag |