missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Wee1, GST Tag Recombinant Protein
WEE1 is classified as a Ser/Thr kinase by sequence analysis.
Brand: Invitrogen™ PV3817
Code : 50
Additional Details : Weight : 0.05250kg
Description
WEE1, which is classified as a Ser⁄Thr kinase by sequence analysis, has been shown to inactivate multiple CDKs by tyrosine phosphorylation.Specifications
NP_003381.1 | |
3716, 7465 | |
10 μg | |
WEE Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
10 μg | |
Dry Ice | |
WEE1 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
99.1 kDa | |
WEE1 | |
Human | |
GST Tag | |
Biochemical binding/kinetic | |
Ser⁄Thr Kinases: Other | |
N-Terminal | |
WEE1 homolog | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |