missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Wee1, GST Tag Recombinant Protein

Product Code. 10148672
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 10148672

Brand: Invitrogen™ PV4072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length

WEE1, which is classified as a Ser/Thr kinase by sequence analysis, has been shown to inactivate multiple CDKs by tyrosine phosphorylation. Wee1 protein catalyzes the inhibitory tyrosine phosphorylation of CDC2/cyclin B kinase, and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P30291
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 7465
Molecular Weight (g/mol) 99.1 kDa
Name Human Wee1, GST Tag
Quantity 1 mg
Regulatory Status RUO - research use only
Gene Alias CTRCT30; DKFZp686I18166; epididymis luminal protein 113; fa20h03; FLJ16446; HEL113; VIM; Vimentin; wee 1 homolog; WEE 1 homolog 1 (S. pombe); Wee1; WEE1 G2 checkpoint kinase; WEE1 homolog; wee1 protein; WEE1 tyrosine kinase; WEE1+ homolog; Wee1A; wee1A kinase; WEE1hu; wee1-like protein kinase; wu:fa20h03; zgc:136514
Common Name WEE1
KinaseFamily WEE Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.