missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASHC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£147.00 - £216.00
Specifications
| Antigen | WASHC1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 5-10uL/mg of uLysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
WASHC1 Polyclonal antibody specifically detects WASHC1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| WASHC1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CXYorf1-Like Protein On Chromosome 9, FAM39E, Family With Sequence Similarity 39, Member E, FLJ00075 Protein, Protein FAM39E, WAS Protein Family Homolog 1, WASH, WASH Complex Subunit 1, WASH1 | |
| Produced in rabbits immunized with E. coli-derived Human WASHC1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10uL/mg of uLysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 100287171 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title