missing translation for 'onlineSavingsMsg'
Learn More

WASHC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-12683-100ul

 View more versions of this product

Product Code. 18620278

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

WASHC1 Polyclonal antibody specifically detects WASHC1 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WASHC1
Polyclonal
Western Blot 1:500-1:2000, Immunoprecipitation 5-10uL/mg of uLysate
CXYorf1-Like Protein On Chromosome 9, FAM39E, Family With Sequence Similarity 39, Member E, FLJ00075 Protein, Protein FAM39E, WAS Protein Family Homolog 1, WASH, WASH Complex Subunit 1, WASH1
Produced in rabbits immunized with E. coli-derived Human WASHC1 fragment
100 μg
Primary
Human
Purified
Western Blot, Immunoprecipitation
Unconjugated
PBS, pH7.0
Rabbit
Antigen and protein A Affinity-purified
RUO
100287171
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.