missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASHC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12683-50ul
This item is not returnable.
View return policy
Description
WASHC1 Polyclonal antibody specifically detects WASHC1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| WASHC1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10uL/mg of uLysate | |
| CXYorf1-Like Protein On Chromosome 9, FAM39E, Family With Sequence Similarity 39, Member E, FLJ00075 Protein, Protein FAM39E, WAS Protein Family Homolog 1, WASH, WASH Complex Subunit 1, WASH1 | |
| Produced in rabbits immunized with E. coli-derived Human WASHC1 fragment | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 100287171 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction