Learn More
Waters Corporation Viridis Silica 2-EP Column
Designed and tested for SFC, the Viridis columns demonstrate a wide range of selectivity for greater resolution.
£612.00 - £23700.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 100 Å |
Quantity | 1/Ea. |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10140000
|
Waters Corporation
186004951 |
30 mm | 250 mm |
£9510.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17152962
|
Waters Corporation
186004946 |
19 mm | 250 mm |
£4770.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17122972
|
Waters Corporation
186004954 |
50 mm | 150 mm |
£16200.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17142962
|
Waters Corporation
186004945 |
19 mm | 150 mm |
£3840.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17102972
|
Waters Corporation
186004952 |
50 mm | 50 mm |
£9255.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17172962
|
Waters Corporation
186004948 |
30 mm | 75 mm |
£5085.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17132962
|
Waters Corporation
186004944 |
19 mm | 100 mm |
£3075.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17162962
|
Waters Corporation
186004947 |
30 mm | 50 mm |
£4080.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17112962
|
Waters Corporation
186004937 |
4.6 mm | 150 mm |
£765.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17112972
|
Waters Corporation
186004953 |
50 mm | 100 mm |
£13575.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17182962
|
Waters Corporation
186004949 |
30 mm | 100 mm |
£5700.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
15198648
|
Waters Corporation
186004955 |
50 mm | 250 mm |
£23700.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17143302
|
Waters Corporation
186005800 |
3 mm | 50 mm |
£612.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17153302
|
Waters Corporation
186005801 |
3 mm | 100 mm |
£684.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17163302
|
Waters Corporation
186005802 |
3 mm | 150 mm |
£745.50
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17173302
|
Waters Corporation
186005803 |
3 mm | 250 mm |
£810.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
17193752
|
Waters Corporation
186006542 |
2.1 mm | 150 mm |
£745.50
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
Description
- The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes – without the use of mobile-phase additives – even for well-retained basic achiral compounds.
Specifications
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
Column |
Spherical | |
100 Å | |
No |