missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation Viridis Silica 2-EP Column
Designed and tested for SFC, the Viridis columns demonstrate a wide range of selectivity for greater resolution.
Marke: Waters Corporation 186005800
Weitere Details : Weight : 0.10960kg
Beschreibung
- The reduction and control of surface silanol activity on Viridis Hybrid particles delivers, under SFC conditions, excellent peak shapes – without the use of mobile-phase additives – even for well-retained basic achiral compounds.
Spezifikation
3 mm | |
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
Column |
50 mm | |
Spherical | |
100 Å | |
No |