missing translation for 'onlineSavingsMsg'
Learn More

Villin Ab-1 Mouse Monoclonal Antibody, Epredia™

Villin Ab-1, Mouse Monoclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

£96.85 - £521.00

Specifications

Antigen Villin Ab-1
Clone CWWB1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12639936
View Documents Promotion Details
Epredia
MS-1499-P
Concentrated 1 mL
£521.00
1mL
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12659936
View Documents Promotion Details
Epredia
MS-1499-P1
Concentrated 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12649936
View Documents Promotion Details
Epredia
MS-1499-P0
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12689936
View Documents Promotion Details
Epredia
MS-1499-R7
Prediluted 7 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12699936
View Documents Promotion Details
Epredia
MS-1499-RQ
Ready-to-Use 12 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Villin can cap, nucleate, sever and bundle actin in a Ca and phosphoinositide-regulated manner. It is associated with the microvillar actin core bundle of intestinal and renal brush border implicated in adsorption. Villin is composed of six repeats, each containing 150 residues that together constitute the core domain followed by the carboxyl-terminal headpiece domain of 87 residues. The core domain retains the Ca dependent capping nucleating and severing activity, whereas the headpiece domain contributes towards actin filament bundling and binding F-actin, independently of Ca

Host Species: Mouse

Clone: CWWB1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Immunogen: Human Villin protein

Molecular Weight: 92.5kDa

Positive Control: Small intestine and kidney

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Villin Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human Villin protein
CWWB1
Monoclonal
Mouse
Cytoskeletal Protein
Primary
Videos
SDS
Documents

Documents

Product Certifications