missing translation for 'onlineSavingsMsg'
Learn More

Villin Ab-1 Mouse Monoclonal Antibody, Epredia™

Villin Ab-1, Mouse Monoclonal Ab is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Brand:  Epredia MS-1499-P0

Code : LD

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12649936

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Villin can cap, nucleate, sever and bundle actin in a Ca and phosphoinositide-regulated manner. It is associated with the microvillar actin core bundle of intestinal and renal brush border implicated in adsorption. Villin is composed of six repeats, each containing 150 residues that together constitute the core domain followed by the carboxyl-terminal headpiece domain of 87 residues. The core domain retains the Ca dependent capping nucleating and severing activity, whereas the headpiece domain contributes towards actin filament bundling and binding F-actin, independently of Ca

Host Species: Mouse

Clone: CWWB1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Immunogen: Human Villin protein

Molecular Weight: 92.5kDa

Positive Control: Small intestine and kidney

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Villin Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cytoskeletal Protein
Human
IgG1
Immunohistochemistry (Paraffin)
CWWB1
Unconjugated
Human Villin protein
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers