Learn More
Kartell™ Variable Volume Pluripet Micropipettor
Ideal for dispensing large volumes of rather thick and viscous aqueous fluids. Kartell™ Variable Volume Pluripet Micropipettor is equipped with a special filter that protects the internal piston by increasing its lifetime and preventing contaminations.
£320.00 - £356.00
Specifications
Certifications/Compliance | EN IS 8655 |
---|---|
Autoclavable | Autoclavable |
Type | Micropipette |
Product Code | Brand | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Compatible Tips | Price | Quantity & Availability | ||||
10407912
|
Kartell™
0132910 |
1000 to 10,000 μL | Kartell™ 943 |
£356.00
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
10070782
|
Kartell™
0132210 |
100 to 1000 μL | Kartell™ 961, 941 |
£320.00
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
Description
- The handle connection ensures a more comfortable and non-fatiguing grip
- The volume can be selected by turning the emptying button or turning the cylinder supplied for the purpose
- The large display ensures simple reading reading of the set volume value
- The button of each micropipettor has a specific colour so identification of the usable volume interval is simple
- The tip ejector is adjustable in height to house a large variety of tips and is can be disassembled to simplify cleaning operations
- The recalibration device ensures quick and precise recalibrations
- The robust structure of the PVDF body ensures high mechanical and chemical resistances
- Totally autoclavable at 121°C for 20 minutes
- The materials used for the structure are resistant and durable, and permit exposure to U.V. rays
Specifications
EN IS 8655 | |
Autoclavable | |
Micropipette |