missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | USP14 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16893184
|
Proteintech
14517-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16883184
|
Proteintech
14517-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.Specifications
| USP14 | |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P54578, Q9JMA1 | |
| 291796, 59025, 9097 | |
| USP14 Fusion Protein Ag5979 | |
| Primary | |
| -20°C | |
| USP14 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Deubiquitinating enzyme 14, TGT, Ubiquitin thiolesterase 14, USP14 | |
| Usp14 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title