Learn More
USP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14517-1-AP-20UL
 View more versions of this product
Product Code. 16893184
  • £135.00 / 20µL
Description
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
Specifications
Specifications
USP14
Polyclonal
Unconjugated
USP14
Deubiquitinating enzyme 14, TGT, Ubiquitin thiolesterase 14, USP14
Rabbit
Antigen Affinity Chromatography
RUO
291796, 59025, 9097
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P54578, Q9JMA1
Usp14
USP14 Fusion Protein Ag5979
20 μL
Primary
Rat, Human, Mouse