missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14517-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.Specifications
| USP14 | |
| Polyclonal | |
| Unconjugated | |
| USP14 | |
| Deubiquitinating enzyme 14, TGT, Ubiquitin thiolesterase 14, USP14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291796, 59025, 9097 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P54578, Q9JMA1 | |
| Usp14 | |
| USP14 Fusion Protein Ag5979 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction